H32915.06
H32915.14
H32915.06EA
88.4
EUR
InStock
H32915.06
H32915.14
4-Amino-3-(trifluormethoxy)benzoesäure 98%
4-Amino-3-(trifluoromethoxy)benzoic acid
Purity: 98%
CAS No.: 175278-22-5
Molecular Formula: C8H6F3NO3
Formula Weight: 221.14
MDL No.: MFCD00205143