H33260.MD
H33260.03
H33260.06
H33260.MDEA
32.8
EUR
InStock
H33260.MD
H33260.03
H33260.06
4-Chloro-5-(4-chlorophenyl)thieno[2,3-d]pyrimidine 96%
4-Chloro-5-(4-chlorophenyl)thieno[2,3-d]pyrimidine
Purity: 96%
CAS No.: 331761-46-7
Molecular Formula: C12H6Cl2N2S
Formula Weight: 281.16
Melting Point: 139.5-141°
MDL No.: MFCD01038375