H33642.03
H33642.06
H33642.03EA
59.8
EUR
InStock
H33642.03
H33642.06
4-(Thiophen-2-yl)benzoic acid 96%
4-(Thiophen-2-yl)benzoic acid
Purity: 96%
CAS No.: 29886-62-2
Molecular Formula: C11H8O2S
Formula Weight: 204.25
Melting Point: 243-245°
MDL No.: MFCD00690742