Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

2-Hydroxy-1,4-naphthochinon 97%, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

Achtung

M-Clarity™ Qualitätsstufe = MQ200

H46805-100G H46805-10G H46805-25G
H46805-100GEA 255 EUR InStock
H46805-100G H46805-10G H46805-25G
2-Hydroxy-1,4-naphthochinon 97%, Sigma-Aldrich®
2-Hydroxy-1,4-naphthochinon
2-Hydroxy-1,4-naphthoquinone is one among the forensic reagents used for fingerprint detection. 2-Hydroxy-1,4-naphthoquinone, produced from Lawsonia inermis, is an orange dye. It belongs to the class of naphthoquinone dyes. 2-Hydroxy-1,4-naphthoquinone is used to dye hair and to color textiles. It has antibacterial, antifungal, anti-inflammatory, antiviral and antineoplastic properties. 2-Hydroxy-1,4-naphthoquinone inhibits tumor cell growth. It stimulates the production of reactive oxygen species (ROS).

  • 2-Hydroxy-1,4-naphthoquinone has been used as a compound for the culture of schistosome worms


Formel: C₁₀H₆O₃
MW: 174.16 g/mol
Melting Pt: 192...195 °C (Zers., Lit.)
Storage Temperature: Room Temperature
MDL Number: MFCD00001678
CAS-Nummer: 83-72-7
EINECS: 201-496-3
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9275e4df331e069b9d0211ca937f12707edc47ee&vid=9275e4df331e069b9d0211ca937f12707edc47ee&prid=web&clid=SAPDAM
Order Now
Spezifikation Testergebnisse
Appearance (colour) Faint yellow to dark yellow and faint orange to dark orange
Appearance (form) Powder
Purity (HPLC area %) ≥96.5 %
Infrared spectrum Conforms to structure

 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge