H50042.03
H50042.MD
H50042.03EA
328
EUR
InStock
H50042.03
H50042.MD
6-Bromo[1,8]naphthyridin-2(1H)-one 98%
6-Bromo[1,8]naphthyridin-2(1H)-one
Purity: 98%
CAS No.: 72754-05-3
Molecular Formula: C8H5BrN2O
Formula Weight: 225.05
Melting Point: 300°
MDL No.: MFCD07781151