H50044.03
H50044.MD
H50044.03EA
121
EUR
InStock
H50044.03
H50044.MD
7-Azaindole-5-boronic acid pinacol ester 97%
7-Azaindole-5-boronic acid pinacol ester
Purity: 97%
CAS No.: 754214-56-7
Molecular Formula: C13H17BN2O2
Formula Weight: 244.10
Melting Point: 241-245°
MDL No.: MFCD08060937