H51058.03
H51058.06
H51058.03EA
157
EUR
InStock
H51058.03
H51058.06
1-Chlor-4-ethinylbenzol ≥98%
1-Chlor-4-ethinylbenzol
Purity: 98%
CAS No.: 873-73-4
Molecular Formula: C8H5Cl
Formula Weight: 136.58
Flash Point: 57°(135°F)
Melting Point: 43-47°
Boiling Point: 179°
UN No.: 1325
MDL No.: MFCD00191917