H51063.MD
H51063.03
H51063.06
H51063.MDEA
56
EUR
InStock
H51063.MD
H51063.03
H51063.06
1,10-Diaza-18-krone-6 (1,4,10,13-Tetraoxa-7,16-diazacyclooctadecan, Kryptofix® 22) 96%
1,10-Diaza-18-krone-6 (1,4,10,13-Tetraoxa-7,16-diazacyclooctadecan, Kryptofix® 22)
Purity: 96%
CAS No.: 23978-55-4
Molecular Formula: C12H26N2O4
Formula Weight: 262.35
Beilstein Reference: 609764
MDL No.: MFCD00005112