H51528.MD
H51528.03
H51528.MDEA
48.5
EUR
InStock
H51528.MD
H51528.03
2-Methoxy-6-(tributylstannyl)pyrazin 95%
2-Methoxy-6-(tributylstannyl)pyrazin
Purity: 95%
CAS No.: 1105511-66-7
Molecular Formula: C17H32N2OSn
Formula Weight: 399.15
Storage Temperature: 2°C to 8°C