H51778.06
H51778.14
H51778.06EA
67.7
EUR
InStock
H51778.06
H51778.14
2-(4-Pyridyl)benzimidazole 97%
2-(4-Pyridyl)benzimidazole
Purity: 97%
CAS No.: 2208-59-5
Molecular Formula: C12H9N3
Formula Weight: 195.22
Odour: Odourless
Melting Point: 216-221°
MDL No.: MFCD00453948