H52407.03
H52407.06
H52407.03EA
114
EUR
InStock
H52407.03
H52407.06
2-Brom-1-methylimidazol 95%
2-Bromo-1-methylimidazole
Purity: 95%
CAS No.: 16681-59-7
Molecular Formula: C4H5BrN2
Formula Weight: 161.00
Flash Point: 110°(230°F)
Boiling Point: 172°
MDL No.: MFCD02179525