H52556.03
H52556.06
H52556.03EA
69.2
EUR
InStock
H52556.03
H52556.06
(R)-1-Boc-3-hydroxypiperidin 97
(R)-1-Boc-3-hydroxypiperidin
Purity: 97%
CAS No.: 143900-43-0
Molecular Formula: C10H19NO3
Formula Weight: 201.27
Flash Point: 104°(219°F)
Melting Point: 46-50°
Optical Rotation: -23° (c=5 in methanol)
MDL No.: MFCD04115306