H53102.MD
H53102.03
H53102.MDEA
74.8
EUR
InStock
H53102.MD
H53102.03
3-Carbamoyl-4-fluorobenzeneboronic acid 97%
3-Carbamoyl-4-fluorobenzeneboronic acid
Purity: 97%
CAS No.: 874219-34-8
Molecular Formula: C7H7BFNO3
Formula Weight: 182.95
Melting Point: 257-259°
MDL No.: MFCD08235053