H53141.MD
H53141.03
H53141.MDEA
70.3
EUR
InStock
H53141.MD
H53141.03
3-(Methylcarbamoyl)phenylboronsäure ≥98%
3-(Methylcarbamoyl)phenylboronsäure
Purity: 98%
CAS No.: 832695-88-2
Molecular Formula: C8H10BNO3
Formula Weight: 178.98
Melting Point: 114-124°
MDL No.: MFCD04038918