H54372.MD
H54372.03
H54372.MDEA
192
EUR
InStock
H54372.MD
H54372.03
2-Amino-4-(trifluormethyl)-5-pyridinboronsäurepinakolester 95%
2-Amino-4-(trifluormethyl)-5-pyridinboronsäurepinakolester
Purity: 95%
CAS No.: 944401-57-4
Molecular Formula: C12H16BF3N2O2
Formula Weight: 288.07
Beilstein Reference: 15519102
MDL No.: MFCD12923420