H55007.03
H55007.06
H55007.03EA
65.3
EUR
InStock
H55007.03
H55007.06
2-Amino-4-chlorpyrimidin 98%
2-Amino-4-chloropyrimidine
Purity: 98%
CAS No.: 3993-78-0
Molecular Formula: C4H4ClN3
Formula Weight: 129.55
Storage Temperature: 2°C to 8°C
Melting Point: 155-160° dec.
MDL No.: MFCD00038021