H55215.06
H55215.14
H55215.06EA
38.6
EUR
InStock
H55215.06
H55215.14
2-(Methylamino)acetonitrile hydrochloride 98%
2-(Methylamino)acetonitrile hydrochloride
Purity: 99%
CAS No.: 25808-30-4
Molecular Formula: C3H6N2.HCl
Formula Weight: 106.55
Melting Point: 103-106°
Beilstein Reference: 3908975
MDL No.: MFCD00012587