H55482.03
H55482.06
H55482.03EA
29.9
EUR
InStock
H55482.03
H55482.06
Methyl-2-amino-3-thiophencarboxylat 97%
Methyl-2-amino-3-thiophencarboxylat
Purity: 97%
CAS No.: 4651-81-4
Molecular Formula: C6H7NO2S
Formula Weight: 157.19
Melting Point: 76-81°
MDL No.: MFCD00159547