H55761.03
H55761.06
H55761.14
H55761.03EA
54.6
EUR
InStock
H55761.03
H55761.06
H55761.14
2,6-Di-tert-butylpyridin ≥97%
2,6-Di-tert-butylpyridin
Purity: 97+%
CAS No.: 585-48-8
Molecular Formula: C13H21N
Formula Weight: 191.32
Flash Point: 71°(160°F)
Refractive Index: 1.4730
Boiling Point: 100-101°/23mm
Beilstein Reference: 125886
Merck Reference: 14,3036
MDL No.: MFCD00006306