H61399.06
H61399.14
H61399.22
H61399.06EA
33.1
EUR
InStock
H61399.06
H61399.14
H61399.22
α,3-Dibrom-4-fluortoluol ≥97%
α,3-Dibrom-4-fluortoluol
Purity: 97%
CAS No.: 78239-71-1
Molecular Formula: C7H5Br2F
Formula Weight: 267.92
Physical Form: Solid
Appearance: White
Melting Point: 41°
UN No.: 3261
MDL No.: MFCD09037367