H61464.AA
H61464.AAEA
70.8
EUR
InStock
H61464.AA
Trimethylboroxine 50% (w/w) in THF ≥98%
Trimethylboroxine
CAS No.: 823-96-1
Molecular Formula: C3H9B3O3
Formula Weight: 125.54
Physical Form: Liquid
Sensitivity: Moisture Sensitive; Hygroscopic
Appearance: Colourless to yellow
Beilstein Reference: 1757008
UN No.: UN2924
MDL No.: MFCD00013354