H64067.03
H64067.06
H64067.03EA
155
EUR
InStock
H64067.03
H64067.06
2,2-dimethylbut-3-enoic acid 95%
2,2-dimethylbut-3-enoic acid
Purity: 95%
CAS No.: 10276-09-2
Molecular Formula: C6H10O2
Formula Weight: 114.14
Storage Temperature: 2°C to 8°C
Physical Form: Liquid
Density: 0.957
Refractive Index: 1.4295
Appearance: Colourless
Melting Point: -6°
Boiling Point: 185°
Beilstein Reference: 1700758
UN No.: UN3265
MDL No.: MFCD00196967