H64394.03
H64394.06
H64394.14
H64394.03EA
33.9
EUR
InStock
H64394.03
H64394.06
H64394.14
2-Chlor-3-nitro-5-(trifluormethyl)pyridin 95%
2-Chloro-3-nitro-5-(trifluoromethyl)pyridine
Purity: 95%
CAS No.: 72587-15-6
Molecular Formula: C6H2ClF3N2O2
Formula Weight: 226.54
Physical Form: Liquid
Appearance: Colourless to pale yellow
Beilstein Reference: 7586778
MDL No.: MFCD06255171