H64485.06
H64485.14
H64485.22
H64485.06EA
37.2
EUR
InStock
H64485.06
H64485.14
H64485.22
8-Azabicyclo[3.2.1]octan-3-onhydrochlorid ≥98%
Nortropinone hydrochloride
Purity: 98+%
CAS No.: 25602-68-0
Molecular Formula: C7H11NO.HCl
Formula Weight: 161.63
Storage Temperature: 2°C to 8°C
Physical Form: Solid
Sensitivity: Hygroscopic
Appearance: White to yellow-brown
Melting Point: 194-196°
Beilstein Reference: 3693524
MDL No.: MFCD03613582