H64774.06
H64774.14
H64774.06EA
73.6
EUR
InStock
H64774.06
H64774.14
3-Oxocyclobutancarbonsäure 98%
3-Oxocyclobutanecarboxylic acid
Purity: 98%
CAS No.: 23761-23-1
Molecular Formula: C5H6O3
Formula Weight: 114.10
Physical Form: Solid
Appearance: Off-white
Melting Point: 69-70°
Beilstein Reference: 2078401
MDL No.: MFCD00100900