H64887.03
H64887.06
H64887.14
H64887.03EA
37.1
EUR
InStock
H64887.03
H64887.06
H64887.14
4-(N,N-Diphenylamino)benzaldehyd 98%
4-(N,N-Diphenylamino)benzaldehyd
Purity: 98%
CAS No.: 4181-05-9
Molecular Formula: C19H15NO
Formula Weight: 273.33
Storage Temperature: 2°C to 8°C
Physical Form: Solid
Appearance: Pale yellow
Odour: Odourless
Melting Point: 129-133° lit.
Beilstein Reference: 2732795
MDL No.: MFCD00145131