Hydrazine colour disc test kit, HY-2
Lieferant: HACH
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
184900
HACH184900EA 245 EUR InStock
HACH184900
Hydrazine colour disc test kit, HY-2
Umwelt-Prüfkits
Proper maintenance and economy of operation of modern boilers and cooling towers require a careful balance between water quality parameters and treatment chemicals. This kit measures hydrazine, a common oxygen scavenger in boiler water, using the simple colour disc method.
Lieferumfang: Includes two 100 ml bottles of deionised water, one reagent, two stoppers, one colour disc comparator, one colour disc, instructions, and a carrying case.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge