Settlometer kit
Lieferant: Hach
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
2115700
HACH2115700EA 309 EUR InStock
HACH2115700
Settlometer kit
Trichter Sedimentiergefäße
This kit is used for analyzing activated sludge process. Custom PP paddle allows agitation of sample without scratching the jar.
  • Easy-to-read white graduations
  • Impact-resistant
  • Chemically resistant at pH range 3 to 10
Lieferumfang: Kit includes a transparent PC 2 liter jar with cover and 10 PC graduated centrifuge tubes (15 ml).
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge