Nitrites test kits, NI-15
Lieferant: HACH
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
2182000
HACH2182000EA 172 EUR InStock
HACH2182000
Nitrites test kits, NI-15
Umwelt-Prüfkits
The accurate visual test kit method to determine nitrogen as nitrite by diazotisation of sample followed by colour comparison.
  • Simple and cost effective
  • Portable test kit
Lieferumfang: Kit includes colour disc, comparator box and tubes, reagents ( but not deionised water), instructions and carrying case.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge