Titrator, digital
Lieferant: Hach
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
2270900
HACH2270900EA 672 EUR InStock
HACH2270900
Titrator, digital
Titratoren
This portable titrator enables analyses in the lab, the plant or the field.
  • Faster than a buret
  • Accuracy ±1%
  • Interchangeable cartridges allow you to change titrants in seconds: No cleaning or rinsing burets
  • More than 50 parameter/range combinations available
  • Concentrated titrants allow 40 to 100 typical titrations per cartridge
  • Number used for calculation of sample concentration is recorded directly from the digital counter
Weight: 132 g
Bestellinformation: Supplied with manual, 100-ml graduated cylinder, 125- and 250- ml Erlenmeyer flasks, demineraliser bottle, clippers, and rugged carrying case.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge