Total hardness reagent set, for digital titrator
Lieferant: HACH
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
2272000
HACH2272000EA 110 EUR InStock
HACH2272000
Total hardness reagent set, for digital titrator
Umwelt-Prüfkits
For total hardness determination by EDTA digital titrator titration with calmagite indicator.
  • Accurate results
  • Fast and flexible
  • Versatile and economical
Digital titrator methods use a high-precision dispensation device and cartridges of concentrated titrant to allow analyses in the lab, the plant or the field. The interchangeable cartridges allow to change titrants in seconds without needing to clean or rinse burettes.
Lieferumfang: Set contains ManVer® 2 indicator, hardness 1 buffer solution and one cartridge each of 0,0800 M and 0,800 M EDTA.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge