Test Kits, model FF-2 for freshwater aquaculture
Lieferant: HACH
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
243001
HACH243001EA 1420 EUR InStock
HACH243001
Test Kits, model FF-2 for freshwater aquaculture
Umwelt-Prüfkits
Maintaining good water quality is essential and this kit utilizes drop count titration and color disc methods to deliver the fast, accurate results. Allows measurement of ten critical freshwater parameters using accurate Digital Titrator titration and continuous colour disc colorimetry.
Lieferumfang: Includes 100 ml bottles of deionised water, twenty three reagents, one flask, clippers, one graduated cylinder, one color comparator box, one digital titrator, one delivery tube, two plastic tubes, one thermometer, one BOD bottle, three colour discs, a manual, and a carrying case.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge