Ammonia (nitrogen) test strips
Lieferant: HACH
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
2755325
HACH2755325EA 21.2 EUR InStock
HACH2755325
Ammonia (nitrogen) test strips
Umwelt-Prüfkits
Ammonia (Nitrogen) is a product of microbiological decay of plant and animal protein. Its presence in raw surface waters usually indicates domestic or agricultural pollution. Above certain levels, it is toxic to fish.
  • Easy to use, disposable, and inexpensive
  • A great way to obtain quick, quantitative answers in the field or in the lab
  • Parameter: ammonia, nitrogen, low range - As NH₃-N, for freshwater
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge