Bi-metallic dial thermometer
Lieferant: Hach
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
4565500
HACH4565500EA 116 EUR InStock
HACH4565500
Bi-metallic dial thermometer
Thermometer Analoge Thermometer
Bi-metallic dial gauge 0 to 200 °C ±1%. Two degree divisions.
Probe Ø 381 mm.
Verpackung: Plastic carry case with clip.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge