Electrode cleaning solution for proteins/organic samples
Lieferant: HACH
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
C20C370
HACHC20C370EA 51.8 EUR InStock
HACHC20C370
Electrode cleaning solution for proteins/organic samples
Elektrolyte Elektrodenaufbewahrungslösungen
Electrode cleaning solution to remove proteins residues organic contaminations/deposits such as food stuff from pH sensing element.
Formula: PEPSIN in HCl
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge