Kombinierte pH-Elektroden, sensION+
Lieferant: Hach
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
LZW5052T97002 LZW5050T97002 LZW5051T97002
HACHLZW5052T97002EA 387 EUR InStock
HACHLZW5052T97002 HACHLZW5050T97002 HACHLZW5051T97002
Kombinierte pH-Elektroden, sensION+
Elektroden pH-Elektroden
These combination electrodes feature a low maintenance 3-in-1 design for the most challenging applications. Equipped with an integrated Pt1000 temperature sensor they provide stable results with low maintenance requirements.
  • 5050T: Heavy-duty electrode handle design optimised for field calibration and storage as it provides a secure interface between the electrode and calibration/storage tube
  • 5051T/5052T: PP working protector prevents accidental breakage of the electrode
The robust electrodes with heavy-duty handle and durable MP5 connector are dedicated for use with Hach sensION+ handheld pH meters.
Order Now
PRODUKT
TECHNISCHE DATEN
ZUBEHÖR
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge