Electrode cleaning solution, RENOVO.X
Lieferant: HACH
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
S16M002
HACHS16M002EA 27.4 EUR InStock
HACHS16M002
Electrode cleaning solution, RENOVO.X
Elektrolyte Elektrodenaufbewahrungslösungen
This extra strong electrode cleaning solution is ideal for use when stronger cleaning agents are required, for example when the solutions contain protein.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge