Vacuum aspiration systems, AE and AK
Lieferant: HAEP LABOR
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
800014900 800014800 800015400 800015500
HAEP800014900EA 770 EUR InStock
HAEP800014900 HAEP800014800 HAEP800015400 HAEP800015500
Vacuum aspiration systems, AE and AK
Pumpen Vakuum-Absaugsysteme
The simple vacuum safety aspiration system, together with the quick lock adapters, filter, ventilation insert and the silicone tubing forms a complete unit. The comfortable aspiration devices additionally have a bottle holder and handle.
  • System optionally available with 2 or 4-litre bottle
  • Screw cap for the bottles with two quick action couplings that close automatically if the hose connections are disengaged
  • Extra-long (3 m) silicone tubing allows the bottle and the vacuum pump to be placed away from the workbench
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge