Spritzen, Serie 1800 Gastight®
Lieferant: HAMILTON BONADUZ
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
84875 84877 84878 84880 84881 84883 84884 84886 84887 84889 7663-01 7659-01 7662-01 7660-01 7661-01 CAL84887 CAL84877 CAL84875 CAL84881 CAL7659-01 CAL7660-01 CAL7661-01 CAL7662-01
549-1214EA 163 EUR InStock
549-1214 549-1215 549-1216 549-1217 549-1218 549-1219 549-1220 549-1221 549-1222 549-1223 HAMI7663-01 HAMI7659-01 HAMI7662-01 HAMI7660-01 HAMI7661-01 HAMICAL84887 HAMICAL84877 HAMICAL84875 HAMICAL84881 HAMICAL7659-01 HAMICAL7660-01 HAMICAL7661-01 HAMICAL7662-01
Spritzen, Serie 1800 Gastight®
Spritzen Spritzen für die Chromatographie
The 1800 series is the reinforced plunger version of the 1700 series syringe. Attached to the flange of the syringe is a removable blue syringe holder that prevents heat transfer, dispense inaccuracies and plunger blow outs. It also provides support to small volume plungers that are prone to bending during injection.
  • Reinforced, protected plunger
  • Ideal for dispensing both liquids and gases in volumes from 1 up to 500 µl
  • Gas sterilisable
  • Stainless steel, borosilicate glass and PTFE fluid path
They have a precision machined PTFE plunger tip which creates a leak-free seal. With the tight fit, the tip essentially wipes the interior of the syringe barrel free of sample. This feature is particularly useful with heterogeneous samples as it reduces the chance that a deposit will occur and cause the plunger to freeze.
Cemented needle (N): Cemented needles for low volume syringes are cemented into the glass syringe barrel at a point corresponding to the zero graduation mark. With this termination, dead volume is limited to the internal volume of the needle. Not autoclavable. Needle gauge is determined by the syringe volume and are not user-selectable.
Removable needle (RN): The design allows the needles to seat precisely at the zero graduation mark of the syringe. Users can select the needle gauge, length, and point style to optimize the syringe for custom applications. This termination also allows for a removable needle without increasing the dead volume of the syringe and is ideal when there is a risk of the needle clogging. Autoclavable when disassembled. Repeated autoclaving will shorten syringe life.
Needle point style 2: Sharp, beveled, curved, non-coring needle point recommended for septum penetration.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge