Digital stopwatch, Profil 25
Lieferant: HANHART
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
225.1721-00
HANH225.1721-00EA 110 EUR InStock
HANH225.1721-00
Digital stopwatch, Profil 25
Timer Stoppuhren, digital
Basic function stopwatch with two-button operation.
  • Display range: 9 hours, 59 minutes, 59,99 seconds
  • Measuring range: 1/100 seconds
  • Start/Stop/Reset – Addition/Split/Lap
  • Time of day
  • 25 memories: 25 split times and 25 lap times
  • Function window for Split, Lap and normal times
  • Robust, water-protected ABS case and a clearly readable LCD display
Weight: 90 g
Lieferumfang: Supplied with protective pouch, cord and battery (AA, 1,5 V).
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge