Syringes
Lieferant: HARVARD APPARATUS
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
70-2256 70-2259
HARV70-2256EA 0 EUR InStock
HARV70-2256 HARV70-2259
Syringes
Spritzen Spritzen, medizinisch
These stainless steel syringes are intended for high pressure applications with good resistance to most aggressive liquids.
  • Wetted parts are made of stainless steel 316 or Viton® and both syringe barrel and plungers are of stainless steel 316
  • 517 bar peak pressure and guaranteed to be leak free for pressures up to 48,3 bar
  • Fits most Harvard pumps
  • Electron beam welded
  • SWAGELOK® fitting for dead volume
Syringes are available with removable replaceable tips. SWAGELOK® syringe to tube fittings are available in different sizes. All tips are interchangeable with all syringes in the series.
All syringes are supplied with inside diameter dimensions for use with microprocessor controlled pumps and rate charts for use with older 'classic' pumps. Replacement Viton O-rings are available, as are the more chemically resistant Chemraz® O-rings.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge