Peristaltic Pump, P-70
Lieferant: HARVARD APPARATUS
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
707000
HARV707000EA 10690 EUR InStock
HARV707000
Peristaltic Pump, P-70
Pumpen Peristaltikpumpen
Peristaltic Pump P-70 offers unparalleled accuracy, reproducibility, and ease of use. P-70 drive provides flow rates from 0,001 to 70 ml/min.
  • Footswitch control; Analog control
  • Volume dispense - dispenses a fixed volume
  • Timed dispense - dispenses for a fixed time at a set flow rate
  • Pump mode - runs continuously at a set flow rate
  • LCD color touch screen with intuitive icon interface for unparalleled ease-of-use
  • Remote design ideal for work inside incubators and fume hoods
  • Interchangeable pump heads
Peristaltic Pump consists of a control unit, a motor drive, a tubing cassette and some sample tubing.
A complete range of programmable functions allow the pump to be easily adapted to a wide range of dispensing applications. The pump has pre-programmed flow profiles for:
-Constant flow
-Flow ramps
-Pulsatile flow
-Concentration based fluid delivery
The ability to separate the motor drive from the controller to facilitate use and save space in incubators and fume hoods. A library of tubing sizes is stored in the pump's memory minimising set up time. Custom tubing can be used allowing complete flexibility. A full range of interchangeable motor drives to allow for economical ease of use over a broad flow rate range.
Control box dimensions: 130×96×207 mm
Pump head dimensions: 254×118×115 mm
Zertifizierungen: ETL (UL, CSA), WEEE, EU RoHS and CB Scheme Approved.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge