These ALPINE air jet test sieves are suitable for fully automatic sieve recognition. These test sieves are made of brass/stainless steel frame with metal wire cloth made from stainless steel.
Zertifizierungen: According to EN ISO/IEC 17025, DIN EN 10204.
Lieferumfang: Supplied with serial number, inspection certificate and calibration certificate on request.