Phlebotomy/Sample Collection Tray
Lieferant: HEATHROW
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
120261
HEAT120261EA 202 EUR InStock
HEAT120261
Phlebotomy/Sample Collection Tray
Schalen/Schüsseln/Töpfe Blutentnahmetabletts
Organise your phlebotomy, I.V. start or catheter insertion supplies with this spacious, robust and easy to clean carry tray.
  • Lightweight easy-to-clean ABS construction
  • Comfort grip on durable acetal plastic handle tested to hold up to 10 kg
  • Comes with a 17 mm well, 60-place, tube rack
Customise the compartments to suit your requirements with the dividers provided.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge