Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Labels, laser Cryo-Babies® and Cryotags®

Lieferant: HEATHROW

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

HEALCRY-1700 HEALCRY-1200
HEATHEALCRY-1700EA 190 EUR InStock
HEATHEALCRY-1700 HEATHEALCRY-1200
Labels, laser Cryo-Babies® and Cryotags®
Etiketten
These labels are for secure uncompromised labelling.

  • Temperature resistance from –196 to 150 °C
  • Liquid and vapor phase LN2, water baths to 100 °C, and autoclaving and moderate ovens to 150 °C
  • Chemically inert, and resistant to most solvents and caustic agents
  • Adheres to most plastics (100% virgin PP tubes recommended), glass, and metals without cracking, peeling, or degrading
  • Label sheets will not jam printers
  • Flexible polyolefin material and acrylic adhesive
  • Unique combo expands and contracts together with temperature fluctuations
  • Autoclavable and disposable

Laser Cryo tags withstand both long term cryogenic storage conditions and conventional freezer storage. Cryogenic labels with a solvent-resistant adhesive that will adhere to croygenic vials, microtubes, test tubes, glass vials, beakers, and metal racks. Laser Cryo Tags also withstand flash/snap freezing in liquid nitrogen, boiling water baths at 100 °C, and dry heat up to 150 °C.

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge