Spatula
Lieferant: HEATHROW
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
HS15909
HEATHS15909EA 9.4 EUR InStock
HEATHS15909
Spatula
Spatel
This spatula is used to transfer, lift, and handle powders and granular materials.
  • Made from nickel-plated, stainless steel wire
  • Autoclavable
Mirror finish with flat and spoon ends.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge