Racks for cryogenic vials
Lieferant: HEATHROW
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
HS23229B
HEATHS23229BEA 53 EUR InStock
HEATHS23229B
Racks for cryogenic vials
Gestelle Gestelle für Kryoröhrchen
Racks to store and transport cryogenic vials.
  • Embossed index numbers and squared, upper left corner aid orientation
  • Ribbed grips make handling with gloves easier
  • Made from PC, can withstand both cryogenic freezing and autoclaving
Well tabs allow one-handed opening and closing of vials that have a slot feature.
Well Ø: 12.9 mm.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge