Cover glasses for hemacytometers, Assistent®
Lieferant: GLASWARENFABRIK KARL HECHT
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
40415710 40416710 40412710 40417710
HECH40415710EA 13.1 EUR InStock
HECH40415710 HECH40416710 HECH40412710 HECH40417710
Cover glasses for hemacytometers, Assistent®
Deckgläser
Assistent® counting chambers are precision items in special optical glass used to count the particles per volume unit of a liquid.
  • Lateral fields for inscription separated by grooves from the three ground and polished supports
  • Central support deeper than the outer ones and referred to as depth of chamber
  • Central support of those chambers divided by an additional groove, coated by a rhodium layer
Zertifizierungen: IVD directive 98/79/EC
Order Now
PRODUKT
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge