Pipette aid, Micro-Pipex
Lieferant: GLASWARENFABRIK KARL HECHT
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
40558001
HECH40558001EA 49.8 EUR InStock
HECH40558001
Pipette aid, Micro-Pipex
Pipetten Pipettierhelfer
Aid for pipetting liquids by using disposable micropipettes with ring mark as well as other pipettes up to 1 ml, e.g. RBC and WBC pipettes.
  • Contaminated disposable micropipettes up to 50 µl are released contactless by the integrated ejection
  • Autoclavable at +121 °C
  • Pipette holder: max. 5 mm Ø
Please note: The pipetting aid may not be used for capillaries end-to-end.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge