Draining rack
Lieferant: GLASWARENFABRIK KARL HECHT
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
41238010
HECH41238010EA 41.9 EUR InStock
HECH41238010
Draining rack
Gestelle Abtropfgestelle
Drying rack made of Plexiglas®, for 12 microscope slides.
The microscope slides are placed vertically on replaceable blotting paper. Two packs of paper can be stored in the base of the drying rack.
Lieferumfang: Supplied with two packs of paper.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge